Learn More
ACROBiosystems Human Cardiotrophin 1 Protein, His Tag (MALS verified)
Catalog No. 502719904
Encompass_Preferred
The protein encoded by this gene is a secreted cytokine that induces cardiac myocyte hypertrophy in vitro. It has been shown to bind and activate the ILST/gp130 receoptor. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-271-9904 Supplier ACROBiosystems Supplier No. CA1H5243100UG
The protein encoded by this gene is a secreted cytokine that induces cardiac myocyte hypertrophy in vitro. It has been shown to bind and activate the ILST/gp130 receoptor. Two transcript variants encoding different isoforms have been found for this gene.