Learn More
Bioworld HUMAN CARDIOTROPHIN-1 (RECO
Supplier:  Bioworld 220602751
Encompass_Preferred
HUMAN CARDIOTROPHIN-1 (RECOMBINANT): Recombinant Human Cardiotrophin-1 (CT-1) Source: E.coli Introduction: Cardiotrophin-1 (CT-1) is a 201 amino acid member of the interleukin-6 superfamily. It was identified by its ability to induce...
Catalog No. 50-253-374