Learn More
Biomatik Corporation Human Cardiotrophin Like Cytokine Factor 1 (CLCF1) CLIA Kit
Supplier:  Biomatik Corporation EKU09822
Encompass_Preferred
Human Cardiotrophin Like Cytokine Factor 1 (CLCF1) CLIA Kit for detecting CLCF1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 5.5-4,000pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9UBD9. Gene ID: 23529. Target Synonyms: CLC; NR6; BSF3; CISS2; NNT1; Novel Neurotrophin-1; B Cell-Stimulating Factor-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8426