Learn More
Biomatik Corporation Human Cardiotrophin Like Cytokine Factor 1 (CLCF1) ELISA Kit
Supplier:  Biomatik Corporation EKU02955
Encompass_Preferred
Human Cardiotrophin Like Cytokine Factor 1 (CLCF1) ELISA Kit for detecting CLCF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBD9. Gene ID: 23529. Target Synonyms: CLC; NR6; BSF3; CISS2; NNT1; Novel Neurotrophin-1; B Cell-Stimulating Factor-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5401