Learn More
American Research Products Inc Human cardiotrophln 1 CT 1 EL
Catalog No. 5000326694
Encompass
Human cardiotrophln 1 CT-1 ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26694 Supplier American Research Products Inc Supplier No. CSBE08454H
Human cardiotrophln 1 CT-1 ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich