Learn More
Biomatik Corporation Human Carnitine Palmitoyltransferase 1A, Liver (CPT1A) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10526
Encompass_Preferred
Human Carnitine Palmitoyltransferase 1A, Liver (CPT1A) Wide-range ELISA Kit for detecting CPT1A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50416. Gene ID: 1374. Target Synonyms: CPT1-L; Carnitine O-palmitoyltransferase I, liver isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6985