Learn More
Biomatik Corporation Human Cartilage Intermediate Layer Protein 2 (CILP2) CLIA Kit
Supplier:  Biomatik Corporation EKU11648
Encompass_Preferred
Human Cartilage Intermediate Layer Protein 2 (CILP2) CLIA Kit for detecting CILP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q8IUL8. Gene ID: 148113. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4753