Learn More
Biomatik Corporation Human Cartilage Oligomeric Matrix Protein (COMP) CLIA Kit
Supplier:  Biomatik Corporation EKU09498
Encompass_Preferred
Human Cartilage Oligomeric Matrix Protein (COMP) CLIA Kit for detecting COMP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.41-300ng/ml. Sensitivity: 0.17ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P49747. Gene ID: 1311. Target Synonyms: MED; EDM1; EPD1; PSACH; THBS5; TSP-5; Pseudoachondroplasia,Epiphyseal Dysplasia 1,Multiple; Thrombospondin 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5707