Learn More
Bioworld HUMAN CASEIN KINASE 2 BETA
Supplier:  Bioworld 220602771
Encompass_Preferred
HUMAN CASEIN KINASE 2 BETA ACTIVE (RECOMBINANT): Recombinant Human Casein Kinase 2 beta (CK2-beta) Source: E.Coli Description : Recombinant Human Casein Kinase 2 beta is a non-glycosylated polypeptide having a molecular mass of 24.9 ...
Catalog No. 50-253-376