Learn More
Bioworld HUMAN CASEIN KINASE 2 HOLOE
Supplier:  Bioworld 220602781
Encompass_Preferred
HUMAN CASEIN KINASE 2 HOLOENZYME (RECOMBINANT): Recombinant Human Casein Kinase 2 Holoenzyme Source: E.Coli Description : Recombinant Human Casein Kinase 2 Holoenzyme is a non-glycosylated polypeptide having a molecular mass of 140 k...
Catalog No. 50-253-377