Learn More
Sino Biological Recombinant Anti-Caspase-14 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11856R145100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Caspase-14 / CASP14 (rh Caspase-14 / CASP14; Catalog#11856-H07E; NP_036246.1; Ser 2-Gln 242).
Catalog No. 50-255-4500