Learn More
Sino Biological Recombinant Anti-Caspase-14 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 11856R006P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Caspase-14 / CASP14 (rh Caspase-14 / CASP14; Catalog#11856-H07E; NP_036246.1; Ser 2-Gln 242) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4502