Learn More
Sino Biological Anti-Caspase-14 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11856RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Caspase-14 / CASP14 (rh CASP14; Catalog#11856-H07E; NP_036246.1; Ser2-Gln242). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4499