Learn More
Sino Biological Human CASP3 PAb 20ul (Sino Biological)
Supplier:  Sino Biological 100561T3220
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human CASP3, and purified by antigen affinity chromatography.
Catalog No. 50-237-2024