Learn More
Sino Biological Anti-Caspase-9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 310365T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CASP9 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-8454