Learn More
Biomatik Corporation Human Caspase 14 (CASP14) CLIA Kit
Supplier:  Biomatik Corporation EKU09426
Encompass_Preferred
Human Caspase 14 (CASP14) CLIA Kit for detecting CASP14 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.56pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P31944. Target Synonyms: Caspase 14 Apoptosis-Related Cysteine Peptidase; Cysteinyl Aspartate Specific Proteinases 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7765