Learn More
Biomatik Corporation Human Caspase 2 (CASP2) ELISA Kit
Supplier:  Biomatik Corporation EKU12491
Encompass_Preferred
Human Caspase 2 (CASP2) ELISA Kit for detecting CASP2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42575. Gene ID: 835. Target Synonyms: ICH1L; NEDD2; ICH-1L/1S; Cysteinyl Aspartate Specific Proteinases 2; Apoptosis-Related Cysteine Peptidase; Neural Precursor Cell Expressed Developmentally Down-Regulated 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7955