Learn More
Cusabio Technology LLC Human Caspase 3Casp-3 ELISA
Catalog No. 5000049290
Encompass_Preferred
This kit detects CASP3 (Human Caspase 3) in serum plasma cell lysates and tissue homogenates with a sensitivity of 0 312 ng/mL and a detection range of 0 312 20 ng/mL Utilizing a sandwich ELISA methodology it provides quantitative results with a lower limit of detection at 0 078 ng/mL ensuring precise measurement of low-abundance targets The assay requires 50 100 L of sample per test and can be completed within 1 5 hours Absorbance is measured at 450 nm offering a reliable and standardized readout Designed for flexibility across multiple sample types this kit is suitable for apoptosis-related research and applications requiring accurate quantification of active Caspase-3 levels in biological matrices
Catalog No. 50-000-49290 Supplier Cusabio Technology LLC Supplier No. CSBE08856H
This kit detects CASP3 (Human Caspase 3) in serum plasma cell lysates and tissue homogenates with a sensitivity of 0 312 ng/mL and a detection range of 0 312 20 ng/mL Utilizing a sandwich ELISA methodology it provides quantitative results with a lower limit of detection at 0 078 ng/mL ensuring precise measurement of low-abundance targets The assay requires 50 100 L of sample per test and can be completed within 1 5 hours Absorbance is measured at 450 nm offering a reliable and standardized readout Designed for flexibility across multiple sample types this kit is suitable for apoptosis-related research and applications requiring accurate quantification of active Caspase-3 levels in biological matrices