Learn More
Abcam Human Caspase 4 ELISA Kit, 1 x 96 Tests
Catalog No. 502655901
Encompass_Preferred
Human Caspase 4 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Caspase 4 with a sensitivity of 6.7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-5901 Supplier Abcam Supplier No. AB2750981X96TESTS
Human Caspase 4 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Caspase 4 with a sensitivity of 6.7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement