Learn More
Biomatik Corporation Human Caspase 6 (CASP6) ELISA Kit
Supplier:  Biomatik Corporation EKU02994
Encompass_Preferred
Human Caspase 6 (CASP6) ELISA Kit for detecting CASP6 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55212. Gene ID: 839. Target Synonyms: MCH2; Cysteinyl Aspartate Specific Proteinases 6; Caspase 6,Apoptosis-Related Cysteine Peptidase; Apoptotic protease Mch-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9071