Learn More
American Research Products Inc Human Caspase 9 Casp 9 ELISA
Catalog No. 5000326744
Encompass
Human Caspase-9 Casp-9 ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26744 Supplier American Research Products Inc Supplier No. CSBE08862H
Human Caspase-9 Casp-9 ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich