Learn More
ACROBiosystems Human Catenin beta-1 Protein, His Tag (MALS verified)
Catalog No. 502331883
Encompass_Preferred
Human Catenin beta-1 Protein, His Tag (CA1-H51H3) is expressed from E.coli cells. It contains AA Asn 138 - Glu 686 (Accession # P35222-1).
Catalog No. 50-233-1883 Supplier ACROBiosystems Supplier No. CA1H51H3100UG
Human Catenin beta-1 Protein, His Tag (CA1-H51H3) is expressed from E.coli cells. It contains AA Asn 138 - Glu 686 (Accession # P35222-1).