Learn More
ACROBiosystems Human Catenin beta-1 25ug
Catalog No. 502535447
Encompass_Preferred
Biotinylated Human Catenin beta-1 HisAvitag (CA1-H81E3) is expressed from E. coli cells. It contains AA Asn 138 - Glu 686 (Accession # P35222-1).
Catalog No. 50-253-5447 Supplier ACROBiosystems Supplier No. CA1H81E325UG
Biotinylated Human Catenin beta-1 HisAvitag (CA1-H81E3) is expressed from E. coli cells. It contains AA Asn 138 - Glu 686 (Accession # P35222-1).