Learn More
Sino Biological Recombinant Human Cystatin C Protein (His Tag)
Supplier:  Sino Biological 10439H08H100
Encompass_Preferred
A DNA sequence encoding the human Cystatin-C (NP_000090.1) (Ser 27-Ala 146) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6894