Learn More
Sino Biological Recombinant Human Cathepsin C Protein (His Tag)
Supplier:  Sino Biological 10484H08H50
Encompass_Preferred
A DNA sequence encoding the pro form of human cathepsin C (NP_001805.3) (Met 1-Leu 463) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6895