Learn More
Abcam Human Cathepsin D ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1195861X96TESTS
Encompass_Preferred
Human Cathepsin D ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Cathepsin D in Human in Plasma, Tissue Extracts, Cell culture supernatant, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1480287