Learn More
Biomatik Corporation Human Cathepsin E (CTSE) ELISA Kit
Supplier:  Biomatik Corporation EKU03029
Encompass_Preferred
Human Cathepsin E (CTSE) ELISA Kit for detecting CTSE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14091. Gene ID: 1510. Target Synonyms: CTS-E; CATE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5176