Learn More
Abcam Human Cathepsin G ELISA Kit, 96TEST
Supplier:  Abcam AB28527796TEST
Encompass_Preferred
Human Cathepsin G ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Cathepsin G in Human in Plasma, Tissue Homogenate, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5464