Learn More
Raybiotech Inc Human Cathepsin H ELISA Kit
Supplier:  Raybiotech Inc ELHCATHEPSINH1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.17 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Cathepsin H.
Catalog No. 50-194-9065