Learn More
ACROBiosystems Human Cathepsin L 50ug
Supplier:  ACROBiosystems CALH52H350UG
Encompass_Preferred
Human Cathepsin L Protein His Tag (CAL-H52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 18 - Val 333 (Accession # P07711-1).
Catalog No. 50-253-5416