Learn More
Sino Biological Recombinant Human Cathepsin L Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10486H08H50
Encompass_Preferred
A DNA sequence encoding the pro form of human Cathepsin-L1 (NP_001903.1) (Met 1-Val 333) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7138
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'