Learn More
Sino Biological Recombinant Human Cathepsin S Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10487H08H50
Encompass_Preferred
A DNA sequence encoding the pro form of human CTSS (NP_004070.3) (Met 1-Ile 331) with a carboxy-terminal polyhistidine tag was expressed.
Catalog No. 50-224-7051