Learn More
Biomatik Corporation Human Cathepsin Z (CTSZ) ELISA Kit
Supplier:  Biomatik Corporation EKU03044
Encompass_Preferred
Human Cathepsin Z (CTSZ) ELISA Kit for detecting CTSZ in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBR2. Gene ID: 1522. Target Synonyms: CTS-Z; CTSX; Cathepsin P; Cathepsin X. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6958