Learn More
Sino Biological Human CAV2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 106821T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CAV2/Caveolin-2, and purified by antigen affinity chromatography.
Catalog No. 50-237-3568