Learn More
Sino Biological Anti-CBFB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14569RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CBFB / CBF-beta (rh CBFB; Catalog#14569-H07E; Q13951-1; Met1-Pro182). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4126