Learn More
Biomatik Corporation Human Cbp/p300 Interacting Transactivator, With Glu/Asp Rich Carboxy Terminal Domain 1 (CITED1) ELISA Kit
Supplier:  Biomatik Corporation EKU11811
Encompass_Preferred
Human Cbp/p300 Interacting Transactivator, With Glu/Asp Rich Carboxy Terminal Domain 1 (CITED1) ELISA Kit for detecting CITED1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99966. Gene ID: 4435. Target Synonyms: MSG1; Melanocyte-specific protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6199