Learn More
Sino Biological Anti-CBR3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14632RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CBR3 / HCBR3 / Carbonyl Reductase 3 (rh CBR3 / HCBR3 / Carbonyl Reductase 3; Catalog#14632-H07E; O75828; Met1-Trp277). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4123