Learn More
BIOHIPPO Human CC16 Microsample
SDP
Catalog No. 5000315768
Encompass
Human CC16 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CC16 in serum plasma tissue homogenates cell lysates lung lavage fluid cell culture supernates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 11 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CC16 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CC16 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CC16 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15768 Supplier BIOHIPPO Supplier No. BHE15212651
Human CC16 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CC16 in serum plasma tissue homogenates cell lysates lung lavage fluid cell culture supernates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 11 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CC16 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CC16 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CC16 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color