Learn More
Sino Biological Recombinant Human CCK4/PTK7 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 19399H08H100
Encompass_Preferred
A DNA sequence encoding the human PTK7 (NP_002812.2) (Met1-Thr704) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7027