Learn More
ACROBiosystems Biotinylated Human CCL1, His,Avitag (CC1-H82E9) is expressed from human 293 cells (HEK293). It contains AA Lys 24 - Lys 96 (Accession # P22362-1).
Catalog No. 502253237
Encompass_Preferred
Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. CCL1, a member of the CC subfamily, is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine (C-C motif) receptor 8.
This item is not returnable. View return policy
Catalog No. 50-225-3237 Supplier ACROBiosystems Supplier No. CC1H82E9100UG
Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. CCL1, a member of the CC subfamily, is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine (C-C motif) receptor 8.