Learn More
Kingfisher Biotech Inc Human EGF ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1595H004
Encompass_Preferred
The Human EGF ELISA contains capture antibody, standard, and detection antibody for development of a Human EGF ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Human EGF-A ELISA can also be used to measure Chimpanzee, Pygmy Chimpanzee, and Western Lowlands Gorilla EGF. Alternate names include EGF, HOMG4, URG, epidermal growth factor, Epidermal growth factor. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5544