Learn More
Sino Biological Human TGF-beta 3 / TGFB3 Protein (Fc Tag) 100ug
Supplier:  Sino Biological 10434H01H100
Encompass_Preferred
A DNA sequence encoding the human TGFB3 (P10600-1) (Ala301-Ser412) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-222-2707