Learn More
Sino Biological Anti-CCL14 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10476MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCL14 (rh CCL14; Catalog#10476-H07E; Q16627-1; Thr20-Asn93). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1573