Learn More
Sino Biological Human CCL14 Matched ELISA Antibody Pair Set 15 Plates
Supplier: Sino Biological SEK1047615
The Human CCL14 ELISA Pair Set is for the quantitative determination of Human CCL14.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CCL14 coated on a 96-well plate. Standards and samples are added to the wells, and any Human CCL14 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human CCL14 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CCL14 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.