Learn More
Sino Biological Anti-CCL14 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10476T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CCL14 (rh CCL14; Catalog#10476-H07E; Q16627-1; Thr20-Asn93). CCL14 specific IgG was purified by Human CCL14 affinity chromatography.
Catalog No. 50-240-9793