Learn More
Sino Biological Recombinant Human CCL14 Protein (His Tag)
Supplier:  Sino Biological 10476H07E1
Encompass_Preferred
A DNA sequence encoding the mature form of human CCL14 (Q16627-1) (Thr 20-Asn 93) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-255-5282