Learn More
Sino Biological Human CCL15 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10898RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CCL15 / MIP-5 / MIP-1 delta (rh CCL15 / MIP-5 / MIP-1 delta; Catalog#10898-H08E; Q16663-1; Gln 22-Ile 113). CCL15 / MIP-5 / MIP-1 delta specific IgG was purified by Human CCL15 / MIP-5 / MIP-1 delta affinity chromatography.
Catalog No. 50-237-2988