Learn More
Sino Biological Anti-LEC/CCL16 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10491MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CCL16 / HCC-4 / NCC4 (rh CCL16 / HCC-4 / NCC4; Catalog#10491-H07E; NP_004581.1; Gln24-Gln120). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8782