Learn More
Sino Biological Anti-LEC/CCL16 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10491T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CCL16 / HCC-4 / NCC4 (rh CCL16 / HCC-4 / NCC4; Catalog#10491-H07E; NP_004581.1; Gln24-Gln120). CCL16 / HCC-4 / NCC4 specific IgG was purified by Human CCL16 / HCC-4 / NCC4 affinity chromatography.
Catalog No. 50-241-1162