Learn More
ACROBiosystems Human CCL17, Fc Tag (CC7-H5256) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 94 (Accession # Q92583-1).
Supplier:  ACROBiosystems CC7H525620UG
Encompass_Preferred
This antimicrobial gene is one of several Cys-Cys (CC) cytokine genes clustered on the q arm of chromosome 16. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for T lymphocytes, but not monocytes or granulocytes. The product of this gene binds to chemokine receptors CCR4 and CCR8. This chemokine plays important roles in T cell development in thymus as well as in trafficking and activation of mature T cells.
Catalog No. 50-225-3812